Search
Search

MMACHC| Gene Symbol: MMACHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266340.1 | NP_001253269.1 | 199 | 66 | ||
| GenBank mRNA | JU323187.1 | - | 169 | 66 | ||
| JU475075.1 | - | 199 | 66 | |||
Set Membership: |
|