Search
Search

CTNNBL1| Gene Symbol: CTNNBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261665.1 | NP_001248594.1 | 390 | 68 | ||
| GenBank mRNA | JU320149.1 | - | 393 | 68 | ||
| JU473649.1 | - | 390 | 68 | |||
Set Membership: |
|