Search
Search

PCSK4| Gene Symbol: PCSK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122355.2 | XP_014977841.1 | 1674 | 103 | ||
| XM_028838635.1 | XP_028694468.1 | 1776 | 103 | |||
| XM_028838636.1 | XP_028694469.1 | 1749 | 103 | |||
| XM_028838637.1 | XP_028694470.1 | 1749 | 103 | |||
| XM_028838638.1 | XP_028694471.1 | 1701 | 103 | |||
| XM_028838639.1 | XP_028694472.1 | 1515 | 103 | |||
| XM_028838640.1 | XP_028694473.1 | 1487 | 103 | |||
| XM_028838641.1 | XP_028694474.1 | 1882 | 103 | |||
| XM_028838642.1 | XP_028694475.1 | 1563 | 103 | |||
| GenBank mRNA | JV654472.1 | - | 179 | 103 | ||
Set Membership: |
|