Search
Search

ZFYVE1| Gene Symbol: ZFYVE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144034.2 | XP_014999520.1 | 278 | 63 | ||
| XM_015144035.2 | XP_014999521.1 | 340 | 63 | |||
| XM_015144036.2 | XP_014999522.1 | 285 | 63 | |||
| XM_028851649.1 | XP_028707482.1 | 2131 | 63 | |||
| GenBank mRNA | JU322078.1 | - | 2027 | 63 | ||
| JU472360.1 | - | 2076 | 63 | |||
| JV043250.1 | - | 2067 | 63 | |||
| JV638710.1 | - | 1964 | 63 | |||
Set Membership: |
|