Search
Search

| Gene Symbol: REPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137349.2 | XP_014992835.1 | 2386 | 66 | ||
| XM_015137350.2 | XP_014992836.1 | 2383 | 66 | |||
| XM_015137351.2 | XP_014992837.1 | 2383 | 66 | |||
| XM_015137352.2 | XP_014992838.1 | 2323 | 66 | |||
| XM_015137354.2 | XP_014992840.1 | 2305 | 66 | |||
| XM_015137355.2 | XP_014992841.1 | 2302 | 66 | |||
| XM_028847162.1 | XP_028702995.1 | 2380 | 66 | |||
| XM_028847163.1 | XP_028702996.1 | 2320 | 66 | |||
| GenBank mRNA | JU321713.1 | - | 1860 | 66 | ||
| JU336818.1 | - | 1941 | 66 | |||
| JV043406.1 | - | 1896 | 66 | |||
| JV048504.1 | - | 1977 | 66 | |||
| Gene Symbol: REXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV066609.1 | - | 995 | 66 | ||
Set Membership: |
|