Search
Search

U2SURP| Gene Symbol: U2SURP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843754.1 | XP_028699587.1 | 953 | 109 | ||
| XM_028843755.1 | XP_028699588.1 | 950 | 109 | |||
| XM_028843756.1 | XP_028699589.1 | 953 | 109 | |||
| XM_028843757.1 | XP_028699590.1 | 1775 | 109 | |||
| GenBank mRNA | JU333656.1 | - | 864 | 109 | ||
| JU474907.1 | - | 864 | 109 | |||
| JV046571.1 | - | 880 | 109 | |||
| JV489161.1 | - | 867 | 109 | |||
| JV646707.1 | - | 923 | 109 | |||
Set Membership: |
|