Search
Search

CDK15| Gene Symbol: CDK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110669.2 | XP_014966155.1 | 658 | 78 | ||
| XM_015110670.2 | XP_014966156.1 | 655 | 78 | |||
| XM_015110671.2 | XP_014966157.1 | 1439 | 78 | |||
| XM_015110672.2 | XP_014966158.1 | 751 | 78 | |||
| XM_015110673.2 | XP_014966159.1 | 407 | 78 | |||
| XM_028830981.1 | XP_028686814.1 | 754 | 78 | |||
| XM_028830982.1 | XP_028686815.1 | 409 | 78 | |||
Set Membership: |
|