Search
Search

DNAH2| Gene Symbol: DNAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118524.2 | XP_014974010.1 | 3974 | 86 | ||
| XM_015118525.2 | XP_014974011.1 | 3974 | 86 | |||
| XM_015118526.2 | XP_014974012.1 | 3974 | 86 | |||
| XM_028835972.1 | XP_028691805.1 | 3974 | 86 | |||
Set Membership: |
|