Search
Search

SLC11A1| Gene Symbol: SLC11A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111049.2 | XP_014966535.2 | 1627 | 69 | ||
| XM_015111050.2 | XP_014966536.1 | 1556 | 69 | |||
| XM_028831104.1 | XP_028686937.1 | 1436 | 69 | |||
| XM_028831105.1 | XP_028686938.1 | 1433 | 69 | |||
| XM_028831106.1 | XP_028686939.1 | 1313 | 69 | |||
| XM_028831107.1 | XP_028686940.1 | 1507 | 69 | |||
Set Membership: |
|