Search
Search

PEX6| Gene Symbol: PEX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089644.4 | XP_001089644.2 | 2570 | 73 | ||
| XM_015136401.2 | XP_014991887.2 | 2570 | 73 | |||
| GenBank mRNA | JU322602.1 | - | 2309 | 73 | ||
| JU488114.1 | - | 2039 | 73 | |||
| JV564988.1 | - | 2348 | 73 | |||
| JV637054.1 | - | 2368 | 73 | |||
Set Membership: |
|