Search
Search

ZNF292| Gene Symbol: ZNF292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136906.2 | XP_014992392.1 | 8699 | 98 | ||
| XM_028847378.1 | XP_028703211.1 | 7005 | 98 | |||
| XM_028847379.1 | XP_028703212.1 | 6935 | 98 | |||
| XM_028847380.1 | XP_028703213.1 | 6394 | 98 | |||
| GenBank mRNA | JU333520.1 | - | 6249 | 98 | ||
| JU474789.1 | - | 6870 | 98 | |||
| U63016.1 | - | 532 | 98 | |||
Set Membership: |
|