Search
Search

PRDX3| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265826.2 | NP_001252755.1 | 463 | 71 | ||
| GenBank mRNA | JU326514.1 | - | 278 | 71 | ||
| JU479186.1 | - | 390 | 71 | |||
| JV600249.1 | - | 463 | 71 | |||
Set Membership: |
|