Search
Search

SMIM29| Gene Symbol: SMIM29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002803700.3 | XP_002803746.3 | 347 | 144 | ||
| XM_015136124.2 | XP_014991610.2 | 596 | 144 | |||
| XM_015136125.2 | XP_014991611.2 | 301 | 144 | |||
| GenBank mRNA | JU333405.1 | - | 359 | 144 | ||
| JU333406.1 | - | 478 | 144 | |||
| JU474692.1 | - | 212 | 144 | |||
| JV046454.1 | - | 372 | 144 | |||
| JV608948.1 | - | 354 | 144 | |||
Set Membership: |
|