Search
Search

ARG2| Gene Symbol: ARG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261253.1 | NP_001248182.1 | 936 | 96 | ||
| XM_015143926.2 | XP_014999412.1 | 1298 | 96 | |||
| GenBank mRNA | JU321469.1 | - | 908 | 96 | ||
| JU321470.1 | - | 908 | 96 | |||
| JU473538.1 | - | 936 | 96 | |||
| JU473539.1 | - | 936 | 96 | |||
| JU473540.1 | - | 936 | 96 | |||
| JU473541.1 | - | 936 | 96 | |||
| JV565398.1 | - | 960 | 96 | |||
Set Membership: |
|