Search
Search

SFXN5| Gene Symbol: SFXN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103924.4 | XP_001103924.1 | 1062 | 111 | ||
| XM_015112647.2 | XP_014968133.1 | 1062 | 111 | |||
| XM_028832380.1 | XP_028688213.1 | 993 | 111 | |||
| XM_028832381.1 | XP_028688214.1 | 1062 | 111 | |||
| XM_028832382.1 | XP_028688215.1 | 1301 | 111 | |||
| XM_028832383.1 | XP_028688216.1 | 104 | 111 | |||
| GenBank mRNA | JV633841.1 | - | 530 | 111 | ||
| JV665953.1 | - | 558 | 111 | |||
Set Membership: |
|