Search
Search

C6H5orf22| Gene Symbol: C6H5orf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193783.2 | NP_001180712.2 | 492 | 165 | ||
| XM_015139778.2 | XP_014995264.2 | 493 | 165 | |||
| XM_028849085.1 | XP_028704918.1 | 493 | 165 | |||
| XM_028849086.1 | XP_028704919.1 | 229 | 165 | |||
| GenBank mRNA | JU335319.1 | - | 453 | 165 | ||
| JU476273.1 | - | 466 | 165 | |||
| JV512018.1 | - | 470 | 165 | |||
Set Membership: |
|