Search
Search

PALB2| Gene Symbol: PALB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125798.2 | XP_014981284.2 | 2786 | 79 | ||
| XM_015125799.2 | XP_014981285.2 | 2786 | 79 | |||
| GenBank mRNA | JU348564.1 | - | 2500 | 79 | ||
| JV048268.1 | - | 2603 | 79 | |||
Set Membership: |
|