Search
Search

TMEM237| Gene Symbol: TMEM237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257889.1 | NP_001244818.1 | 949 | 98 | ||
| XM_015110633.2 | XP_014966119.2 | 918 | 98 | |||
| XM_028830481.1 | XP_028686314.1 | 899 | 98 | |||
| XM_028830482.1 | XP_028686315.1 | 846 | 98 | |||
| GenBank mRNA | JU335979.1 | - | 949 | 98 | ||
| JU335980.1 | - | 891 | 98 | |||