Search
Search

LOC694255| Gene Symbol: LOC694255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837186.1 | XP_028693019.1 | 603 | 97 | ||
| XM_028837187.1 | XP_028693020.1 | 597 | 97 | |||
| XM_028837188.1 | XP_028693021.1 | 495 | 97 | |||
| XM_028837189.1 | XP_028693022.1 | 1315 | 97 | |||
Set Membership: |
|