Search
Search

FERMT3| Gene Symbol: FERMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113898.2 | XP_014969384.1 | 1637 | 62 | ||
| XM_015113900.2 | XP_014969386.1 | 1636 | 62 | |||
| GenBank mRNA | JU471543.1 | - | 1226 | 62 | ||
| JU494553.1 | - | 600 | 62 | |||
Set Membership: |
|