Search
Search

PCNX3| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113776.2 | XP_014969262.1 | 5526 | 63 | ||
| XM_015113777.2 | XP_014969263.1 | 5523 | 63 | |||
| XM_015113778.2 | XP_014969264.1 | 5523 | 63 | |||
| XM_015113779.2 | XP_014969265.1 | 5508 | 63 | |||
| XM_015113780.2 | XP_014969266.1 | 5496 | 63 | |||
| XM_015113781.2 | XP_014969267.1 | 5487 | 63 | |||
| XM_028833233.1 | XP_028689066.1 | 5526 | 63 | |||
| GenBank mRNA | JU343452.1 | - | 4916 | 63 | ||
| JU475576.1 | - | 4963 | 63 | |||
Set Membership: |
|