Search
Search

CDK18| Gene Symbol: CDK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852445.1 | XP_028708278.1 | 2886 | 77 | ||
| XM_028852450.1 | XP_028708283.1 | 2792 | 77 | |||
| XM_028852451.1 | XP_028708284.1 | 1105 | 77 | |||
| XM_028852454.1 | XP_028708287.1 | 962 | 77 | |||
| GenBank mRNA | JU334846.1 | - | 999 | 77 | ||
| JU334847.1 | - | 1089 | 77 | |||
| JV601604.1 | - | 1043 | 77 | |||
| JV668792.1 | - | 1021 | 77 | |||
Set Membership: |
|