Search
Search

RECK| Gene Symbol: RECK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083599.4 | XP_001083599.1 | 1519 | 116 | ||
| XM_015117456.2 | XP_014972942.1 | 1519 | 116 | |||
| XM_015117457.2 | XP_014972943.1 | 1661 | 116 | |||
| XM_015117458.2 | XP_014972944.1 | 1561 | 116 | |||
| XM_028835061.1 | XP_028690894.1 | 1271 | 116 | |||
| GenBank mRNA | JU494835.1 | - | 1379 | 116 | ||
| JV044926.1 | - | 1490 | 116 | |||
| JV635472.1 | - | 1508 | 116 | |||
| JV725076.1 | - | 1349 | 116 | |||
Set Membership: |
|