Search
Search

SLC16A6| Gene Symbol: SLC16A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808134.3 | XP_002808180.2 | 745 | 69 | ||
| XM_015120178.2 | XP_014975664.1 | 533 | 69 | |||
| GenBank mRNA | JU496088.1 | - | 372 | 69 | ||
Set Membership: |
|