Search
Search

MAP3K8| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146718.2 | XP_015002204.1 | 1232 | 114 | ||
| XM_015146719.2 | XP_015002205.1 | 2479 | 114 | |||
| XM_015146720.2 | XP_015002206.1 | 1246 | 114 | |||
| XM_028826131.1 | XP_028681964.1 | 1105 | 114 | |||
| GenBank mRNA | JV642458.1 | - | 890 | 114 | ||
Set Membership: |
|