Search
Search

PI4K2B| Gene Symbol: PI4K2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808407.3 | XP_002808453.2 | 215 | 114 | ||
| XM_015138118.2 | XP_014993604.1 | 215 | 114 | |||
| GenBank mRNA | JU489228.1 | - | 120 | 114 | ||
Set Membership: |
|