Search
Search

DPY19L1| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845871.1 | XP_028701704.1 | 1320 | 102 | ||
| XM_028845872.1 | XP_028701705.1 | 1164 | 102 | |||
| XM_028845873.1 | XP_028701706.1 | 1235 | 102 | |||
| GenBank mRNA | JU327592.1 | - | 937 | 102 | ||
| JV056200.1 | - | 939 | 102 | |||
Set Membership: |
|