Search
Search

BOC| Gene Symbol: BOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129873.2 | XP_014985359.2 | 2675 | 108 | ||
| XM_015129874.2 | XP_014985360.2 | 2533 | 108 | |||
| XM_015129876.2 | XP_014985362.2 | 2666 | 108 | |||
| XM_015129877.2 | XP_014985363.2 | 2423 | 108 | |||
| XM_028844621.1 | XP_028700454.1 | 2524 | 108 | |||
| XM_028844622.1 | XP_028700455.1 | 6327 | 108 | |||
| XM_028844623.1 | XP_028700456.1 | 2530 | 108 | |||
| XM_028844624.1 | XP_028700457.1 | 2533 | 108 | |||
| XM_028844625.1 | XP_028700458.1 | 2389 | 108 | |||
| XM_028844626.1 | XP_028700459.1 | 2076 | 108 | |||
| GenBank mRNA | JV636447.1 | - | 2003 | 108 | ||
Set Membership: |
|