Search
Search

MYO19| Gene Symbol: MYO19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119450.2 | XP_014974936.2 | 1572 | 96 | ||
| GenBank mRNA | JU348830.1 | - | 179 | 96 | ||
| JU477294.1 | - | 1460 | 96 | |||
| JV048302.1 | - | 1571 | 96 | |||
Set Membership: |
|