Search
Search

TCTN2| Gene Symbol: TCTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109635.2 | XP_014965121.1 | 385 | 82 | ||
| XM_015109637.2 | XP_014965123.1 | 385 | 82 | |||
| GenBank mRNA | JU477253.1 | - | 361 | 82 | ||
| JV045680.1 | - | 309 | 82 | |||
| JV635711.1 | - | 374 | 82 | |||