Search
Search

IGFL4| Gene Symbol: IGFL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838679.1 | XP_028694512.1 | 1120 | 60 | ||
| XM_028838680.1 | XP_028694513.1 | 1451 | 60 | |||
| XM_028838681.1 | XP_028694514.1 | 1120 | 60 | |||
| XM_028838682.1 | XP_028694515.1 | 1120 | 60 | |||
| XM_028838683.1 | XP_028694516.1 | 1120 | 60 | |||
| XM_028838684.1 | XP_028694517.1 | 1119 | 60 | |||
| XM_028838685.1 | XP_028694518.1 | 1118 | 60 | |||
| XM_028838686.1 | XP_028694519.1 | 917 | 60 | |||
| GenBank mRNA | JU343874.1 | - | 25 | 60 | ||
Set Membership: |
|