Search
Search

C15H9orf50| Gene Symbol: C15H9orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116639.2 | XP_014972125.2 | 1359 | 69 | ||
| XM_015116640.2 | XP_014972126.2 | 1996 | 69 | |||
| XM_028834732.1 | XP_028690565.1 | 1997 | 69 | |||
| XM_028834733.1 | XP_028690566.1 | 1943 | 69 | |||
| XM_028834734.1 | XP_028690567.1 | 1999 | 69 | |||
Set Membership: |
|