Search
Search

CATSPERB| Gene Symbol: CATSPERB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808468.3 | XP_002808514.3 | 328 | 100 | ||
| XM_028851766.1 | XP_028707599.1 | 307 | 100 | |||
| XM_028851767.1 | XP_028707600.1 | 329 | 100 | |||
Set Membership: |
|