Search
Search
| Gene Symbol: Aldob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012496.2 | NP_036628.2 | 1 - 2 | 42 | 64 | |
| GenBank mRNA | BC081697.1 | - | 1 - 2 | 42 | 7109764 | 64 |
| FQ209538.1 | - | 1 - 2 | 62 | 64 | ||
| FQ209594.1 | - | 1 - 2 | 62 | 64 | ||
| FQ209615.1 | - | 1 - 2 | 62 | 64 | ||
| FQ209621.1 | - | 1 - 2 | 60 | 64 | ||
| FQ209639.1 | - | 1 - 2 | 62 | 64 | ||
| FQ209643.1 | - | 1 - 2 | 59 | 64 | ||
| FQ209782.1 | - | 1 - 2 | 62 | 64 | ||
| FQ209819.1 | - | 1 - 2 | 72 | 64 | ||
| FQ210002.1 | - | 1 - 2 | 62 | 64 | ||
| FQ210849.1 | - | 1 - 2 | 62 | 64 | ||
| FQ211083.1 | - | 1 - 2 | 62 | 64 | ||
| FQ211156.1 | - | 1 - 2 | 62 | 64 | ||
| FQ211159.1 | - | 1 - 2 | 62 | 64 | ||
| FQ218139.1 | - | 1 - 2 | 62 | 64 | ||
| FQ218217.1 | - | 1 - 2 | 62 | 64 | ||
| FQ218348.1 | - | 1 - 2 | 68 | 64 | ||
| FQ218522.1 | - | 1 - 2 | 62 | 64 | ||
| FQ218551.1 | - | 1 - 2 | 62 | 64 | ||
| FQ218665.1 | - | 1 - 2 | 62 | 64 | ||
| FQ218911.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219096.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219116.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219261.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219453.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219555.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219585.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219640.1 | - | 1 - 2 | 60 | 64 | ||
| FQ219646.1 | - | 1 - 2 | 62 | 64 | ||
| FQ219703.1 | - | 1 - 2 | 62 | 64 | ||
| M10149.1 | - | 1 - 2 | 53 | 64 | ||
Set Membership: |
|