Search
Search
| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013033.2 | NP_037165.1 | 8 - 9 | 1058 | 80 | |
| GenBank mRNA | AB000729.1 | - | 8 - 9 | 1084 | 80 | |
| BC081827.1 | - | 8 - 9 | 1090 | 7129017 | 80 | |
| D16101.1 | - | 8 - 9 | 1058 | 80 | ||
| U03120.1 | - | 8 - 9 | 1016 | 80 | ||
Set Membership: |
|