Search
Search
| Gene Symbol: Stx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031124.1 | NP_112386.1 | 3 - 4 | 289 | 61 | |
| XM_006231097.2 | XP_006231159.1 | 3 - 4 | 373 | 61 | ||
| XM_008760262.1 | XP_008758484.1 | 3 - 4 | 373 | 61 | ||
| XM_008760263.1 | XP_008758485.1 | 3 - 4 | 373 | 61 | ||
| XM_008760264.1 | XP_008758486.1 | 3 - 4 | 373 | 61 | ||
| XM_008760265.1 | XP_008758487.1 | 3 - 4 | 373 | 61 | ||
| XM_008760266.1 | XP_008758488.1 | 3 - 4 | 373 | 61 | ||
| XM_008760267.1 | XP_008758489.1 | 3 - 4 | 373 | 61 | ||
| GenBank mRNA | BC168246.1 | - | 3 - 4 | 243 | 5600098 | 61 |
| L20820.1 | - | 3 - 4 | 289 | 61 | ||
Set Membership: |
|