Search
Search
| Gene Symbol: Wipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057192.2 | NP_476540.2 | 3 - 4 | 514 | 105 | |
| GenBank mRNA | AJ303456.1 | - | 2 - 3 | 347 | 105 | |
| BC072475.1 | - | 3 - 4 | 514 | 7100751 | 105 | |
Set Membership: |
|