Search
Search
| Gene Symbol: Itgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012711.1 | NP_036843.1 | 24 - 25 | 2973 | 74 | |
| XM_006230214.2 | XP_006230276.1 | 25 - 26 | 3096 | 74 | ||
| GenBank mRNA | AF268593.1 | - | 24 - 25 | 2973 | 74 | |
Set Membership: |
|