Search
Search
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035255.1 | NP_001030332.1 | 7 - 8 | 881 | 120 | |
| XM_006233540.2 | XP_006233602.1 | 8 - 9 | 921 | 120 | ||
| XM_006233541.2 | XP_006233603.1 | 8 - 9 | 920 | 120 | ||
| XM_006233542.2 | XP_006233604.1 | 8 - 9 | 921 | 120 | ||
| XM_006233543.2 | XP_006233605.1 | 9 - 10 | 978 | 120 | ||
| XM_008761567.1 | XP_008759789.1 | 8 - 9 | 922 | 120 | ||
| XM_008761568.1 | XP_008759790.1 | 9 - 10 | 3969 | 120 | ||
| XM_008761569.1 | XP_008759791.1 | 9 - 10 | 985 | 120 | ||
| XM_008761570.1 | XP_008759792.1 | 5 - 6 | 1169 | 120 | ||
| GenBank mRNA | BC099157.1 | - | 7 - 8 | 881 | 7381485 | 120 |
| FQ209707.1 | - | 8 - 9 | 887 | 120 | ||
| FQ234514.1 | - | 7 - 8 | 896 | 120 | ||
Set Membership: |
|