Search
Search
| Gene Symbol: Aldoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012495.2 | NP_036627.1 | 1 - 2 | 142 | 85 | |
| GenBank mRNA | BC064440.1 | - | 1 - 2 | 119 | 5623628 | 85 |
| FQ212782.1 | - | 1 - 2 | 93 | 85 | ||
| FQ213869.1 | - | 1 - 2 | 123 | 85 | ||
| FQ214123.1 | - | 1 - 2 | 93 | 85 | ||
| FQ214510.1 | - | 1 - 2 | 95 | 85 | ||
| FQ224221.1 | - | 1 - 2 | 133 | 85 | ||
| M12919.1 | - | 1 - 2 | 142 | 85 | ||
Set Membership: |
|