Search
Search

Arfgap2| Gene Symbol: Arfgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033707.1 | NP_001028879.1 | 8 - 9 | 715 | 94 | |
| GenBank mRNA | BC101917.1 | - | 8 - 9 | 715 | 7935829 | 94 |
| FQ218185.1 | - | 8 - 9 | 787 | 94 | ||
Set Membership: |
|