Search
Search
| Gene Symbol: Fgd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107617.2 | NP_001101087.2 | 8 - 9 | 1208 | 87 | |
| XM_006256205.2 | XP_006256267.1 | 8 - 9 | 1254 | 87 | ||
| XM_008772729.1 | XP_008770951.1 | 8 - 9 | 1254 | 87 | ||
| GenBank mRNA | BC089841.1 | - | 8 - 9 | 1236 | 87 | |
Set Membership: |
|