Search
Search
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139042.2 | NP_620611.2 | 15 - 16 | 2513 | 153 | |
| GenBank mRNA | AF024622.1 | - | 15 - 16 | 2513 | 153 | |
| U33847.1 | - | 606 | 153 | |||
Set Membership: |
|