Search
Search
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047103.1 | NP_001040568.1 | 6 - 7 | 781 | 102 | |
| XM_006250323.2 | XP_006250385.1 | 7 - 8 | 1053 | 102 | ||
| GenBank mRNA | BC161811.1 | - | 6 - 7 | 782 | 7317639 | 102 |
| DQ272743.1 | - | 6 - 7 | 781 | 102 | ||
Set Membership: |
|