Search
Search
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108329.1 | NP_001101799.1 | 4 - 5 | 1045 | 157 | |
| XM_006248835.2 | XP_006248897.1 | 4 - 5 | 1172 | 157 | ||
| XM_008768976.1 | XP_008767198.1 | 3 - 4 | 6091 | 157 | ||
Set Membership: |
|