Search
Search
| Gene Symbol: Anxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024155.3 | NP_077069.3 | 3 - 4 | 179 | 64 | |
| XM_006236841.2 | XP_006236903.1 | 4 - 5 | 266 | 64 | ||
| XM_006236842.2 | XP_006236904.1 | 3 - 4 | 802 | 64 | ||
| GenBank mRNA | BC085688.1 | - | 3 - 4 | 179 | 7191458 | 64 |
| D38224.2 | - | 1 - 2 | 88 | 64 | ||
Set Membership: |
|