Search
Search

B3galt4| Gene Symbol: B3galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133553.2 | NP_598237.2 | 1 | 1513 | 129 | |
| GenBank mRNA | AB003478.1 | - | 1 | 1484 | 129 | |
| BC126083.1 | - | 1 | 1513 | 8375818 | 129 | |
Set Membership: |
|