Search
Search
| Gene Symbol: Lcn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106560.1 | NP_001100030.1 | 4 - 5 | 434 | 152 | |
| XM_008761622.1 | XP_008759844.1 | 4 - 5 | 454 | 152 | ||
| GenBank mRNA | DQ537496.1 | - | 4 - 5 | 423 | 152 | |
Set Membership: |
|