Search
Search
| Gene Symbol: Zbtb47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244165.2 | XP_006244227.1 | 2 - 3 | 1680 | 67 | |
| XM_006244166.2 | XP_006244228.1 | 2 - 3 | 1714 | 67 | ||
| XM_006244167.2 | XP_006244229.1 | 2 - 3 | 1687 | 67 | ||
| XM_006244168.2 | XP_006244230.1 | 2 - 3 | 1577 | 67 | ||
| XM_008766708.1 | XP_008764930.1 | 2 - 3 | 1522 | 67 | ||
Set Membership: |
|