Search
Search
| Gene Symbol: Mlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031053.1 | NP_112315.1 | 12 - 13 | 1412 | 155 | |
| XM_008766652.1 | XP_008764874.1 | 10 - 11 | 1174 | 155 | ||
| GenBank mRNA | U80054.1 | - | 12 - 13 | 1412 | 155 | |
Set Membership: |
|